Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TH
Transcript:
NM_199292.2
Assay Name:
Mutation genome location:
chr.11 2168003-2168495 on build GRCh38
Assay ID
Hs00326768_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
synaptic transmission, dopaminergic
response to amphetamine
heart morphogenesis
dopamine biosynthetic process from tyrosine
fatty acid metabolic process
sphingolipid metabolic process
heart development
visual perception
sensory perception of sound
learning
memory
mating behavior
locomotory behavior
regulation of heart contraction
response to water deprivation
response to light stimulus
response to herbicide
response to salt stress
anatomical structure morphogenesis
organ morphogenesis
response to zinc ion
multicellular organism aging
response to activity
aminergic neurotransmitter loading into synaptic vesicle
glycoside metabolic process
phthalate metabolic process
cerebral cortex development
response to nutrient levels
response to estradiol
response to lipopolysaccharide
isoquinoline alkaloid metabolic process
social behavior
cellular response to drug
response to isolation stress
response to immobilization stress
neurotransmitter biosynthetic process
terpene metabolic process
dopamine biosynthetic process
epinephrine biosynthetic process
norepinephrine biosynthetic process
catecholamine biosynthetic process
eye photoreceptor cell development
circadian sleep/wake cycle
eating behavior
response to peptide hormone
pigmentation
response to ethanol
response to ether
response to pyrethroid
embryonic camera-type eye morphogenesis
response to corticosterone
response to electrical stimulus
phytoalexin metabolic process
oxidation-reduction process
cellular response to manganese ion
cellular response to nicotine
cellular response to glucose stimulus
cellular response to growth factor stimulus
tyrosine 3-monooxygenase activity
protein binding
ferrous iron binding
ferric iron binding
amino acid binding
oxygen binding
enzyme binding
protein domain specific binding
tetrahydrobiopterin binding
dopamine binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline