Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PAX6
Transcript:
Assay Name:
Mutation genome location:
chr.11 31784650-31785186 on build GRCh38
Assay ID
Hs00327778_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
establishment of mitotic spindle orientation
blood vessel development
eye development
cell fate determination
neuron migration
negative regulation of protein phosphorylation
positive regulation of neuroblast proliferation
lens development in camera-type eye
type B pancreatic cell differentiation
pancreatic A cell development
transcription from RNA polymerase II promoter
smoothened signaling pathway
axon guidance
central nervous system development
salivary gland morphogenesis
visual perception
response to wounding
regulation of asymmetric cell division
organ morphogenesis
dorsal/ventral axis specification
positive regulation of gene expression
protein ubiquitination
oligodendrocyte cell fate specification
cerebral cortex regionalization
forebrain anterior/posterior pattern specification
forebrain dorsal/ventral pattern formation
commitment of neuronal cell to specific neuron type in forebrain
forebrain-midbrain boundary formation
regulation of transcription from RNA polymerase II promoter involved in spinal cord motor neuron fate specification
regulation of transcription from RNA polymerase II promoter involved in ventral spinal cord interneuron specification
regulation of transcription from RNA polymerase II promoter involved in somatic motor neuron fate commitment
pituitary gland development
signal transduction involved in regulation of gene expression
keratinocyte differentiation
regulation of cell migration
positive regulation of epithelial cell differentiation
lacrimal gland development
protein localization to organelle
eye photoreceptor cell development
glucose homeostasis
negative regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of timing of cell differentiation
embryonic camera-type eye morphogenesis
neuron fate commitment
astrocyte differentiation
negative regulation of epithelial cell proliferation
negative regulation of neurogenesis
retina development in camera-type eye
iris morphogenesis
cornea development in camera-type eye
negative regulation of neural precursor cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
protein binding
transcription factor binding
protein kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
co-SMAD binding
R-SMAD binding
HMG box domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline