Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LOC105369535
Transcript:
NM_003105.5
Assay Name:
Mutation genome location:
chr.11 121478098-121478624 on build GRCh38
Gene:
SORL1
Transcript:
NM_003105.5
Assay Name:
Mutation genome location:
chr.11 121478098-121478624 on build GRCh38
Assay ID
Hs00335130_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
chaperone
apolipoprotein
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to Golgi
protein targeting
protein targeting to lysosome
lipid transport
post-Golgi vesicle-mediated transport
receptor-mediated endocytosis
signal transduction
cholesterol metabolic process
regulation of smooth muscle cell migration
negative regulation of protein binding
negative regulation of protein oligomerization
negative regulation of MAP kinase activity
protein retention in Golgi apparatus
positive regulation of protein catabolic process
negative regulation of neurogenesis
protein maturation
positive regulation of protein exit from endoplasmic reticulum
negative regulation of neuron death
negative regulation of beta-amyloid formation
positive regulation of choline O-acetyltransferase activity
negative regulation of tau-protein kinase activity
positive regulation of ER to Golgi vesicle-mediated transport
positive regulation of early endosome to recycling endosome transport
negative regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
positive regulation of protein localization to early endosome
negative regulation of neurofibrillary tangle assembly
positive regulation of endocytic recycling
beta-amyloid binding
transmembrane signaling receptor activity
protein binding
low-density lipoprotein particle binding
ADP-ribosylation factor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline