Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
DNM1L
Transcript:
NM_012062.4
Assay Name:
Mutation genome location:
chr.12 32731117-32731640 on build GRCh38
Assay ID
Hs00335423_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial fission
release of cytochrome c from mitochondria
dynamin polymerization involved in mitochondrial fission
endocytosis
cellular component disassembly involved in execution phase of apoptosis
regulation of mitochondrion organization
peroxisome fission
regulation of protein oligomerization
positive regulation of apoptotic process
positive regulation of GTPase activity
mitochondrial fragmentation involved in apoptotic process
positive regulation of protein secretion
protein oligomerization
protein homotetramerization
heart contraction
membrane fusion
necroptotic process
mitochondrion morphogenesis
protein localization to mitochondrion
positive regulation of mitochondrial fission
mitochondrial membrane fission
positive regulation of release of cytochrome c from mitochondria
regulation of peroxisome organization
regulation of mitophagy
regulation of ATP metabolic process
positive regulation of intrinsic apoptotic signaling pathway
GTPase activity
GTPase activator activity
protein binding
GTP binding
lipid binding
Rab GTPase binding
GTP-dependent protein binding
ubiquitin protein ligase binding
protein homodimerization activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0