Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CSK
Transcript:
NM_004383.2
Assay Name:
Mutation genome location:
chr.15 74798723-74799226 on build GRCh38
Assay ID
Hs00342167_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytokine production
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
central nervous system development
brain development
negative regulation of cell proliferation
negative regulation of low-density lipoprotein particle clearance
cell migration
cell differentiation
T cell costimulation
negative regulation of interleukin-6 production
negative regulation of kinase activity
adherens junction organization
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
negative regulation of Golgi to plasma membrane protein transport
positive regulation of MAP kinase activity
innate immune response
negative regulation of bone resorption
oligodendrocyte differentiation
negative regulation of phagocytosis
T cell receptor signaling pathway
regulation of Fc receptor mediated stimulatory signaling pathway
negative regulation of ERK1 and ERK2 cascade
cellular response to peptide hormone stimulus
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
protein phosphatase binding
protein kinase A catalytic subunit binding
identical protein binding
metal ion binding
proline-rich region binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline