Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TP53
Transcript:
NM_001126113.2
Assay Name:
Mutation genome location:
chr.17 7673853-7674349 on build GRCh38
Assay ID
Hs00346578_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0