Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ICAM1
Transcript:
NM_000201.2
Assay Name:
Mutation genome location:
chr.19 10283941-10284438 on build GRCh38
Assay ID
Hs00353194_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
regulation of leukocyte mediated cytotoxicity
response to amphetamine
T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
acute inflammatory response to antigenic stimulus
T cell antigen processing and presentation
positive regulation of cellular extravasation
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
cell aging
sensory perception of sound
regulation of cell shape
response to ionizing radiation
response to sulfur dioxide
response to organic cyclic compound
membrane to membrane docking
regulation of cell adhesion
extracellular matrix organization
positive regulation of actin filament polymerization
cellular response to nutrient levels
cell adhesion mediated by integrin
response to gonadotropin
response to drug
response to amino acid
positive regulation of GTPase activity
adhesion of symbiont to host
positive regulation of nitric oxide biosynthetic process
response to ethanol
positive regulation of vasoconstriction
response to copper ion
viral entry into host cell
receptor-mediated virion attachment to host cell
positive regulation of peptidyl-tyrosine phosphorylation
regulation of immune response
leukocyte migration
positive regulation of NF-kappaB transcription factor activity
negative regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to alkaloid
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to hypoxia
establishment of Sertoli cell barrier
regulation of ruffle assembly
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of endothelial cell apoptotic process
virus receptor activity
receptor activity
transmembrane signaling receptor activity
integrin binding
protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline