Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RAC1
Transcript:
NM_006908.4
Assay Name:
Mutation genome location:
chr.7 6399708-6400215 on build GRCh38
Assay ID
Hs00368995_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
auditory receptor cell morphogenesis
mast cell chemotaxis
epithelial cell morphogenesis
movement of cell or subcellular component
inflammatory response
hyperosmotic response
cell adhesion
cell-matrix adhesion
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
axon guidance
blood coagulation
cell proliferation
regulation of cell size
response to wounding
anatomical structure morphogenesis
regulation of hydrogen peroxide metabolic process
positive regulation of lamellipodium assembly
regulation of fibroblast migration
positive regulation of cell-substrate adhesion
regulation of neuron maturation
cerebral cortex radially oriented cell migration
embryonic olfactory bulb interneuron precursor migration
cerebral cortex GABAergic interneuron development
lamellipodium assembly
actin cytoskeleton organization
actin filament polymerization
platelet activation
regulation of cell migration
positive regulation of actin filament polymerization
T cell costimulation
ruffle organization
negative regulation of interleukin-23 production
substrate adhesion-dependent cell spreading
positive regulation of Rho protein signal transduction
intracellular signal transduction
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of apoptotic process
positive regulation of phosphatidylinositol 3-kinase activity
engulfment of apoptotic cell
cell-cell junction organization
bone resorption
positive regulation of DNA replication
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
negative regulation of receptor-mediated endocytosis
anatomical structure arrangement
dendrite morphogenesis
cell motility
homeostasis of number of cells within a tissue
regulation of defense response to virus by virus
regulation of small GTPase mediated signal transduction
positive regulation of stress fiber assembly
localization within membrane
positive regulation of focal adhesion assembly
synaptic transmission, GABAergic
Wnt signaling pathway, planar cell polarity pathway
regulation of respiratory burst
cellular response to mechanical stimulus
semaphorin-plexin signaling pathway
dopaminergic neuron differentiation
protein localization to plasma membrane
positive regulation of neutrophil chemotaxis
cochlea morphogenesis
ruffle assembly
positive regulation of substrate adhesion-dependent cell spreading
GTPase activity
protein binding
GTP binding
Rab GTPase binding
enzyme binding
protein kinase binding
GTP-dependent protein binding
thioesterase binding
histone deacetylase binding
Rho GDP-dissociation inhibitor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline