Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NTRK2
Transcript:
NM_006180.4
Assay Name:
Mutation genome location:
chr.9 84744894-84745394 on build GRCh38
Assay ID
Hs00383945_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
neuron migration
positive regulation of protein phosphorylation
inflammatory response
neuromuscular junction development
aging
learning
long-term memory
circadian rhythm
feeding behavior
positive regulation of cell proliferation
response to light stimulus
positive regulation of gene expression
positive regulation of neuron projection development
response to auditory stimulus
glutamate secretion
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
central nervous system neuron development
cerebral cortex development
neuron differentiation
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
neurotrophin signaling pathway
mechanoreceptor differentiation
regulation of GTPase activity
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
retinal rod cell development
protein autophosphorylation
regulation of neurotransmitter secretion
oligodendrocyte differentiation
peripheral nervous system neuron development
positive regulation of axonogenesis
regulation of dendrite development
regulation of protein kinase B signaling
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
retina development in camera-type eye
long-term synaptic potentiation
cellular response to amino acid stimulus
cellular response to tumor necrosis factor
cellular response to brain-derived neurotrophic factor stimulus
negative regulation of anoikis
ATP binding
receptor tyrosine kinase binding
protein homodimerization activity
neurotrophin binding
brain-derived neurotrophic factor binding
brain-derived neurotrophic factor-activated receptor activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline