Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
IGF2
Transcript:
Assay Name:
Mutation genome location:
chr.11 2132113-2132621 on build GRCh38
Gene:
INS-IGF2
Transcript:
Assay Name:
Mutation genome location:
chr.11 2132113-2132621 on build GRCh38
Assay ID
Hs00405504_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
positive regulation of protein phosphorylation
platelet degranulation
glucose metabolic process
regulation of gene expression by genetic imprinting
regulation of transcription, DNA-templated
multicellular organism development
female pregnancy
memory
positive regulation of cell proliferation
insulin receptor signaling pathway
response to radiation
response to nutrient levels
response to estradiol
response to nicotine
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
wound healing
positive regulation of activated T cell proliferation
response to drug
positive regulation of catalytic activity
positive regulation of MAPK cascade
cellular protein metabolic process
response to ethanol
positive regulation of glycogen biosynthetic process
positive regulation of mitotic nuclear division
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of insulin receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
positive regulation of protein kinase B signaling
cellular response to mechanical stimulus
positive regulation of protein serine/threonine kinase activity
positive regulation of receptor activity
positive regulation of glycogen (starch) synthase activity
insulin receptor binding
insulin-like growth factor receptor binding
hormone activity
protein binding
growth factor activity
receptor activator activity
protein serine/threonine kinase activator activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline