Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NRG1
Transcript:
NM_013962.2
Assay Name:
Mutation genome location:
chr.8 31639945-31640480 on build GRCh38
Assay transcription exon location:
Within Exon 1
Assay ID
Hs00408728_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
cell morphogenesis
startle response
cardiac conduction system development
ventricular trabecula myocardium morphogenesis
cell communication
activation of transmembrane receptor protein tyrosine kinase activity
nervous system development
synapse assembly
peripheral nervous system development
locomotory behavior
regulation of blood pressure
positive regulation of cell proliferation
embryo development
neural crest cell development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
peptidyl-tyrosine phosphorylation
glial cell fate commitment
chemorepulsion involved in interneuron migration from the subpallium to the cortex
mammary gland development
positive regulation of myelination
activation of protein kinase B activity
intracellular signal transduction
ERBB signaling pathway
ERBB2 signaling pathway
wound healing
negative regulation of protein catabolic process
myelination
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
positive regulation of GTPase activity
neurotransmitter receptor metabolic process
positive regulation of axon extension
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of Ras protein signal transduction
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
neuron fate commitment
oligodendrocyte differentiation
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of secretion
positive regulation of striated muscle cell differentiation
positive regulation of protein kinase B signaling
cardiac muscle cell differentiation
cardiac muscle cell myoblast differentiation
endocardial cell differentiation
positive regulation of dendritic spine development
positive regulation of calcineurin-NFAT signaling cascade
positive regulation of protein targeting to mitochondrion
positive regulation of protein localization to cell surface
regulation of cell motility
positive regulation of cardiac muscle cell differentiation
negative regulation of neuron migration
transcription cofactor activity
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
cytokine activity
ErbB-2 class receptor binding
integrin binding
growth factor activity
transmembrane receptor protein tyrosine kinase activator activity
receptor tyrosine kinase binding
ErbB-3 class receptor binding
chemorepellent activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline