Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PSEN1
Transcript:
NM_000021.3
Assay Name:
Mutation genome location:
chr.14 73170547-73171132 on build GRCh38
Assay ID
Hs00424193_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
aspartic protease
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
negative regulation of transcription from RNA polymerase II promoter
activation of MAPKK activity
blood vessel development
cell fate specification
somitogenesis
neuron migration
positive regulation of receptor recycling
heart looping
hematopoietic progenitor cell differentiation
T cell activation involved in immune response
neural retina development
protein glycosylation
membrane protein ectodomain proteolysis
mitochondrial transport
cellular response to DNA damage stimulus
response to oxidative stress
negative regulation of epidermal growth factor-activated receptor activity
Notch signaling pathway
Notch receptor processing
memory
post-embryonic development
protein transport
L-glutamate transport
choline transport
synaptic vesicle targeting
single organismal cell-cell adhesion
protein processing
cerebral cortex cell migration
Cajal-Retzius cell differentiation
dorsal/ventral neural tube patterning
embryonic limb morphogenesis
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
endoplasmic reticulum calcium ion homeostasis
beta-amyloid formation
intracellular signal transduction
regulation of phosphorylation
amyloid precursor protein catabolic process
myeloid dendritic cell differentiation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of catalytic activity
regulation of protein binding
positive regulation of MAP kinase activity
negative regulation of neuron apoptotic process
skin morphogenesis
positive regulation of transcription, DNA-templated
regulation of synaptic plasticity
thymus development
neuron development
skeletal system morphogenesis
brain morphogenesis
epithelial cell proliferation
negative regulation of axonogenesis
positive regulation of coagulation
T cell receptor signaling pathway
neuron apoptotic process
negative regulation of ubiquitin-protein transferase activity
smooth endoplasmic reticulum calcium ion homeostasis
regulation of synaptic transmission, glutamatergic
canonical Wnt signaling pathway
regulation of resting membrane potential
positive regulation of dendritic spine development
calcium ion transmembrane transport
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of apoptotic signaling pathway
endopeptidase activity
aspartic-type endopeptidase activity
calcium channel activity
protein binding
beta-catenin binding
PDZ domain binding
cadherin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline