Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CALM1
Transcript:
NM_006888.4
Assay Name:
Mutation genome location:
chr.14 90403590-90404090 on build GRCh38
Assay ID
Hs00424299_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
MAPK cascade
response to amphetamine
regulation of heart rate
platelet degranulation
detection of calcium ion
glycogen catabolic process
muscle contraction
G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
Wnt signaling pathway, calcium modulating pathway
positive regulation of peptidyl-threonine phosphorylation
negative regulation of peptidyl-threonine phosphorylation
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
substantia nigra development
regulation of rhodopsin mediated signaling pathway
positive regulation of cyclic nucleotide metabolic process
positive regulation of protein autophosphorylation
regulation of cytokinesis
positive regulation of phosphoprotein phosphatase activity
ion transmembrane transport
positive regulation of protein dephosphorylation
Fc-epsilon receptor signaling pathway
positive regulation of DNA binding
positive regulation of GTPase activity
inositol phosphate metabolic process
regulation of nitric-oxide synthase activity
positive regulation of nitric-oxide synthase activity
positive regulation of cyclic-nucleotide phosphodiesterase activity
response to corticosterone
response to calcium ion
regulation of cardiac muscle contraction
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of protein serine/threonine kinase activity
regulation of high voltage-gated calcium channel activity
regulation of cell communication by electrical coupling involved in cardiac conduction
Ras guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
adenylate cyclase binding
inositol-1,4,5-trisphosphate 3-kinase activity
ligand-gated ion channel activity
protein kinase binding
protein domain specific binding
nitric-oxide synthase regulator activity
titin binding
type 3 metabotropic glutamate receptor binding
thioesterase binding
N-terminal myristoylation domain binding
phospholipase binding
protein serine/threonine kinase activator activity
phosphatidylinositol 3-kinase binding
ion channel binding
calcium-dependent protein binding
nitric-oxide synthase binding
protein phosphatase activator activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline