Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LEF1
Transcript:
NM_001130714.2
Assay Name:
Mutation genome location:
chr.4 108070452-108070916 on build GRCh38
Assay ID
Hs00433771_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
patterning of blood vessels
osteoblast differentiation
neural crest cell migration
somitogenesis
kidney development
epithelial to mesenchymal transition
sprouting angiogenesis
transcription from RNA polymerase II promoter
Wnt signaling pathway, calcium modulating pathway
positive regulation of cell proliferation
response to lithium ion
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
Wnt signaling pathway
regulation of striated muscle tissue development
dentate gyrus development
hypothalamus development
forebrain radial glial cell differentiation
forebrain neuroblast division
forebrain neuron differentiation
formation of radial glial scaffolds
regulation of cell-cell adhesion
neutrophil differentiation
positive regulation of cell growth
embryonic limb morphogenesis
positive regulation of cell migration
BMP signaling pathway
positive regulation of granulocyte differentiation
mammary gland development
organ regeneration
negative regulation of interleukin-13 production
negative regulation of interleukin-4 production
negative regulation of interleukin-5 production
T cell receptor V(D)J recombination
B cell proliferation
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA binding
steroid hormone mediated signaling pathway
tongue development
skin development
positive regulation by host of viral transcription
histone H3 acetylation
histone H4 acetylation
T-helper 1 cell differentiation
negative regulation of striated muscle tissue development
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
alpha-beta T cell differentiation
eye pigmentation
paraxial mesoderm formation
muscle fiber development
sensory perception of taste
palate development
anatomical structure regression
canonical Wnt signaling pathway
face morphogenesis
cell chemotaxis
apoptotic process involved in morphogenesis
chorio-allantoic fusion
trachea gland development
cellular response to cytokine stimulus
cellular response to interleukin-4
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
odontoblast differentiation
negative regulation of estrogen receptor binding
negative regulation of canonical Wnt signaling pathway
apoptotic process involved in patterning of blood vessels
beta-catenin-TCF complex assembly
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
beta-catenin binding
transcription factor binding
DNA binding, bending
estrogen receptor activity
estrogen receptor binding
enhancer binding
histone binding
histone deacetylase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
C2H2 zinc finger domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0