Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
INSR
Transcript:
NM_000208.3
Assay Name:
Mutation genome location:
chr.19 7116563-7117153 on build GRCh38
Assay transcription exon location:
Within Exon 22
Assay ID
Hs00440288_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
carbohydrate metabolic process
regulation of transcription, DNA-templated
G-protein coupled receptor signaling pathway
positive regulation of cell proliferation
insulin receptor signaling pathway
epidermis development
male gonad development
response to manganese ion
regulation of hydrogen peroxide metabolic process
positive regulation of glycoprotein biosynthetic process
negative regulation of gene expression
response to activity
peptidyl-tyrosine phosphorylation
transformation of host cell by virus
signal transduction by protein phosphorylation
male sex determination
adrenal gland development
positive regulation of cell migration
exocrine pancreas development
activation of protein kinase activity
activation of protein kinase B activity
response to estradiol
negative regulation of transporter activity
cellular response to insulin stimulus
response to vitamin D
response to testosterone
response to tumor necrosis factor
peptidyl-tyrosine autophosphorylation
glucose homeostasis
positive regulation of MAPK cascade
positive regulation of nitric oxide biosynthetic process
fat cell differentiation
response to ethanol
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
regulation of embryonic development
positive regulation of glucose import
protein autophosphorylation
positive regulation of developmental growth
protein heterotetramerization
response to glucocorticoid
positive regulation of meiotic cell cycle
positive regulation of protein kinase B signaling
positive regulation of respiratory burst
cellular response to growth factor stimulus
regulation of female gonad development
protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
insulin-activated receptor activity
insulin-like growth factor receptor binding
protein binding
ATP binding
GTP binding
protein phosphatase binding
lipoic acid binding
insulin-like growth factor I binding
insulin-like growth factor II binding
protein complex binding
3-phosphoinositide-dependent protein kinase binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
PTB domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline