Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CTNNB1
Transcript:
NM_001904.3
Assay Name:
Mutation genome location:
chr.3 41226947-41227479 on build GRCh38
Assay ID
Hs00467769_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
embryonic axis specification
patterning of blood vessels
branching involved in ureteric bud morphogenesis
in utero embryonic development
gastrulation with mouth forming second
cell fate specification
endodermal cell fate commitment
neuron migration
epithelial to mesenchymal transition
neural plate development
liver development
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
lens morphogenesis in camera-type eye
regulation of secondary heart field cardioblast proliferation
metanephros morphogenesis
negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
transcription, DNA-templated
cell adhesion
cell-matrix adhesion
Wnt signaling pathway, calcium modulating pathway
chemical synaptic transmission
ectoderm development
glial cell fate determination
midgut development
negative regulation of cell proliferation
anterior/posterior axis specification
dorsal/ventral axis specification
proximal/distal pattern formation
positive regulation of epithelial to mesenchymal transition
positive regulation of heparan sulfate proteoglycan biosynthetic process
Schwann cell proliferation
response to activity
Wnt signaling pathway
single organismal cell-cell adhesion
stem cell population maintenance
layer formation in cerebral cortex
central nervous system vasculogenesis
osteoclast differentiation
androgen receptor signaling pathway
male genitalia development
hindbrain development
regulation of centriole-centriole cohesion
pancreas development
hair follicle morphogenesis
regulation of myelination
positive regulation of telomere maintenance via telomerase
negative regulation of chondrocyte differentiation
response to estradiol
positive regulation of type I interferon production
T cell differentiation in thymus
negative regulation of protein sumoylation
response to cytokine
adherens junction assembly
protein localization to cell surface
embryonic heart tube development
genitalia morphogenesis
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
hair cell differentiation
catenin import into nucleus
embryonic skeletal limb joint morphogenesis
regulation of T cell proliferation
odontogenesis of dentin-containing tooth
response to drug
embryonic digit morphogenesis
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of MAPK cascade
positive regulation of neuron apoptotic process
response to estrogen
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
myoblast differentiation
bone resorption
positive regulation of endothelial cell differentiation
positive regulation of osteoblast differentiation
negative regulation of osteoclast differentiation
positive regulation of fibroblast growth factor receptor signaling pathway
regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of mitotic cell cycle, embryonic
response to cadmium ion
chromatin-mediated maintenance of transcription
regulation of fibroblast proliferation
cell maturation
synaptic vesicle transport
thymus development
oocyte development
embryonic foregut morphogenesis
positive regulation of skeletal muscle tissue development
regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
negative regulation of oligodendrocyte differentiation
regulation of neurogenesis
synapse organization
positive regulation of sequence-specific DNA binding transcription factor activity
smooth muscle cell differentiation
positive regulation of muscle cell differentiation
protein heterooligomerization
positive regulation of histone H3-K4 methylation
positive regulation of telomerase activity
oviduct development
canonical Wnt signaling pathway
trachea formation
epithelial tube branching involved in lung morphogenesis
lung cell differentiation
lung-associated mesenchyme development
lung induction
epithelial cell differentiation involved in prostate gland development
positive regulation of epithelial cell proliferation involved in prostate gland development
hair follicle placode formation
mesenchymal cell proliferation involved in lung development
endothelial tube morphogenesis
fungiform papilla formation
canonical Wnt signaling pathway involved in positive regulation of cardiac outflow tract cell proliferation
sympathetic ganglion development
cranial ganglion development
regulation of centromeric sister chromatid cohesion
cellular response to mechanical stimulus
cellular response to lithium ion
cellular response to growth factor stimulus
cellular response to indole-3-methanol
renal vesicle formation
renal inner medulla development
renal outer medulla development
nephron tubule formation
regulation of nephron tubule epithelial cell differentiation
regulation of calcium ion import
negative regulation of neuron death
positive regulation of chromatin-mediated maintenance of transcription
regulation of euchromatin binding
regulation of core promoter binding
beta-catenin-TCF complex assembly
beta-catenin destruction complex disassembly
midbrain dopaminergic neuron differentiation
canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
cellular response to insulin-like growth factor stimulus
embryonic brain development
dorsal root ganglion development
regulation of protein localization to cell surface
positive regulation of determination of dorsal identity
positive regulation of DNA-templated transcription, initiation
negative regulation of apoptotic signaling pathway
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
signal transducer activity
protein binding
protein C-terminus binding
transcription factor binding
enzyme binding
kinase binding
protein kinase binding
protein phosphatase binding
estrogen receptor binding
protein complex binding
ionotropic glutamate receptor binding
nuclear hormone receptor binding
transcription regulatory region DNA binding
ion channel binding
alpha-catenin binding
cadherin binding
SMAD binding
androgen receptor binding
nitric-oxide synthase binding
I-SMAD binding
repressing transcription factor binding
cadherin binding involved in cell-cell adhesion
euchromatin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline