Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CREBBP
Transcript:
Assay Name:
Mutation genome location:
chr.16 3726124-3726399 on build GRCh38
Assay ID
Hs00469090_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
stimulatory C-type lectin receptor signaling pathway
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
protein complex assembly
protein acetylation
signal transduction
Notch signaling pathway
cell proliferation
regulation of smoothened signaling pathway
viral process
histone acetylation
N-terminal peptidyl-lysine acetylation
positive regulation of type I interferon production
cellular response to UV
homeostatic process
embryonic digit morphogenesis
regulation of apoptotic process
cellular lipid metabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
positive regulation of cell adhesion molecule production
regulation of transcription from RNA polymerase II promoter in response to hypoxia
response to interleukin-1
cell-cell adhesion
regulation of cellular response to heat
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of NIK/NF-kappaB signaling
beta-catenin-TCF complex assembly
core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
transcriptional repressor activity, RNA polymerase II transcription factor binding
p53 binding
chromatin binding
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity
peptide N-acetyltransferase activity
peroxisome proliferator activated receptor binding
MRF binding
SMAD binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline