Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
EEF2
Transcript:
NM_001961.3
Assay Name:
Mutation genome location:
chr.19 3982406-3982905 on build GRCh38
Assay transcription exon location:
Overlaps Exon 4 - Exon 5
Gene:
SNORD37
Transcript:
NM_001961.3
Assay Name:
Mutation genome location:
chr.19 3982406-3982905 on build GRCh38
Assay transcription exon location:
Overlaps Exon 4 - Exon 5
Assay ID
Hs00502025_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
translation elongation factor
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
response to ischemia
skeletal muscle contraction
translational elongation
aging
glial cell proliferation
response to fluoxetine
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
response to estradiol
response to endoplasmic reticulum stress
skeletal muscle cell differentiation
response to hydrogen peroxide
response to ethanol
positive regulation of translation
response to folic acid
cell-cell adhesion
cellular response to brain-derived neurotrophic factor stimulus
positive regulation of cytoplasmic translation
p53 binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
5S rRNA binding
protein kinase binding
ribosome binding
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline