Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SRC
Transcript:
Assay Name:
Mutation genome location:
chr.20 37396198-37396370 on build GRCh38
Assay ID
Hs00517518_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
cell cycle
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
central nervous system development
cell proliferation
response to mechanical stimulus
response to virus
response to acidic pH
regulation of epithelial cell migration
positive regulation of epithelial cell migration
positive regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of glucose metabolic process
positive regulation of smooth muscle cell migration
single organismal cell-cell adhesion
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
platelet activation
intracellular estrogen receptor signaling pathway
forebrain development
T cell costimulation
protein destabilization
response to nutrient levels
positive regulation of protein autophosphorylation
activation of protein kinase B activity
negative regulation of telomere maintenance via telomerase
negative regulation of protein homooligomerization
cellular response to insulin stimulus
regulation of intracellular estrogen receptor signaling pathway
positive regulation of integrin activation
adherens junction organization
substrate adhesion-dependent cell spreading
cellular response to reactive oxygen species
intracellular signal transduction
osteoclast development
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
regulation of vascular permeability
stress fiber assembly
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein binding
positive regulation of MAP kinase activity
positive regulation of phosphatidylinositol 3-kinase activity
transcytosis
innate immune response
regulation of bone resorption
bone resorption
positive regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of insulin receptor signaling pathway
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
oogenesis
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
progesterone receptor signaling pathway
leukocyte migration
positive regulation of small GTPase mediated signal transduction
response to mineralocorticoid
response to electrical stimulus
regulation of cell cycle
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
negative regulation of telomerase activity
uterus development
branching involved in mammary gland duct morphogenesis
regulation of cell projection assembly
positive regulation of ERK1 and ERK2 cascade
response to interleukin-1
cellular response to lipopolysaccharide
cellular response to peptide hormone stimulus
cellular response to progesterone stimulus
cellular response to fatty acid
cellular response to hypoxia
cellular response to fluid shear stress
regulation of podosome assembly
positive regulation of podosome assembly
positive regulation of protein serine/threonine kinase activity
angiotensin-activated signaling pathway involved in heart process
positive regulation of canonical Wnt signaling pathway
positive regulation of lamellipodium morphogenesis
positive regulation of DNA biosynthetic process
regulation of early endosome to late endosome transport
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
regulation of caveolin-mediated endocytosis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
SH3/SH2 adaptor activity
protein kinase C binding
receptor binding
insulin receptor binding
integrin binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
enzyme binding
kinase binding
heme binding
estrogen receptor binding
SH2 domain binding
ion channel binding
ephrin receptor binding
phosphoprotein binding
hormone receptor binding
growth factor receptor binding
scaffold protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0