Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
STK11
Transcript:
NM_000455.4
Assay Name:
Mutation genome location:
chr.19 1218361-1218513 on build GRCh38
Assay ID
Hs00518951_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
tissue homeostasis
vasculature development
protein phosphorylation
autophagy
cellular response to DNA damage stimulus
cell cycle arrest
spermatid development
axonogenesis
negative regulation of cell proliferation
response to ionizing radiation
positive regulation of autophagy
establishment of cell polarity
regulation of Wnt signaling pathway
negative regulation of cell growth
positive regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase activity
TCR signalosome assembly
glucose homeostasis
anoikis
positive thymic T cell selection
protein autophosphorylation
regulation of dendrite morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of axonogenesis
T cell receptor signaling pathway
negative regulation of lipid biosynthetic process
Golgi localization
regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to UV-B
positive regulation of protein serine/threonine kinase activity
intrinsic apoptotic signaling pathway by p53 class mediator
dendrite extension
positive regulation of protein localization to nucleus
regulation of signal transduction by p53 class mediator
negative regulation of TORC1 signaling
magnesium ion binding
p53 binding
protein serine/threonine kinase activity
protein binding
ATP binding
LRR domain binding
protein kinase activator activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline