Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NTRK3
Transcript:
NM_001012338.2
Assay Name:
Mutation genome location:
chr.15 87880287-87880453 on build GRCh38
Assay ID
Hs00519237_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
neuron migration
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
heart development
circadian rhythm
positive regulation of cell proliferation
positive regulation of gene expression
peptidyl-tyrosine phosphorylation
modulation by virus of host transcription
positive regulation of cell migration
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
neurotrophin signaling pathway
mechanoreceptor differentiation
positive regulation of apoptotic process
response to ethanol
protein autophosphorylation
ephrin receptor signaling pathway
neuron fate specification
response to axon injury
positive regulation of axon extension involved in regeneration
negative regulation of astrocyte differentiation
positive regulation of positive chemotaxis
response to corticosterone
positive regulation of synapse assembly
negative regulation of cell death
lens fiber cell differentiation
cellular response to retinoic acid
cochlea development
activation of GTPase activity
positive regulation of actin cytoskeleton reorganization
p53 binding
GPI-linked ephrin receptor activity
neurotrophin receptor activity
protein binding
ATP binding
neurotrophin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0