Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PPARG
Transcript:
NM_015869.4
Assay Name:
Mutation genome location:
chr.3 12381337-12381493 on build GRCh38
Assay ID
Hs00521155_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
placenta development
negative regulation of acute inflammatory response
transcription initiation from RNA polymerase II promoter
lipid metabolic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
G-protein coupled receptor signaling pathway
heart development
response to nutrient
regulation of blood pressure
response to cold
response to mechanical stimulus
macrophage derived foam cell differentiation
negative regulation of macrophage derived foam cell differentiation
negative regulation of receptor biosynthetic process
negative regulation of cholesterol storage
negative regulation of sequestering of triglyceride
long-chain fatty acid transport
fatty acid oxidation
monocyte differentiation
negative regulation of cell growth
epithelial cell differentiation
response to caffeine
organ regeneration
response to retinoic acid
cellular response to insulin stimulus
negative regulation of collagen biosynthetic process
response to vitamin A
response to lipid
peroxisome proliferator activated receptor signaling pathway
response to immobilization stress
response to diuretic
glucose homeostasis
response to starvation
regulation of circadian rhythm
lipoprotein transport
steroid hormone mediated signaling pathway
response to estrogen
innate immune response
cell fate commitment
positive regulation of fat cell differentiation
low-density lipoprotein particle receptor biosynthetic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
cell maturation
rhythmic process
negative regulation of smooth muscle cell proliferation
positive regulation of oligodendrocyte differentiation
white fat cell differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
negative regulation of telomerase activity
lipid homeostasis
response to low-density lipoprotein particle
positive regulation of phagocytosis, engulfment
negative regulation of interferon-gamma-mediated signaling pathway
regulation of cholesterol transporter activity
regulation of transcription involved in cell fate commitment
cellular response to retinoic acid
cellular response to vitamin E
cellular response to prostaglandin E stimulus
cellular response to hyperoxia
response to metformin
negative regulation of pancreatic stellate cell proliferation
core promoter sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
prostaglandin receptor activity
protein binding
transcription factor binding
drug binding
zinc ion binding
enzyme binding
protein phosphatase binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
activating transcription factor binding
identical protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
retinoid X receptor binding
arachidonic acid binding
alpha-actinin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline