Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Gene:
          PHOX2B
          Transcript:
          NM_003924.3
          Assay Name:
          Mutation genome location:
          chr.4 41747510-41747635 on build GRCh38
          Assay ID Hs00521472_CE

          Purification
          Size
          Availability Made To Order
          Catalog # A15629
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details


          Back To Top

          More Information


          Set Membership:

          Less than 200 bp

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of respiratory gaseous exchange by neurological system process
          noradrenergic neuron differentiation
          noradrenergic neuron development
          brainstem development
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          glial cell differentiation
          regulation of gene expression
          positive regulation of G2/M transition of mitotic cell cycle
          cell differentiation in hindbrain
          medullary reticular formation development
          hindbrain tangential cell migration
          skeletal muscle cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          autonomic nervous system development
          enteric nervous system development
          sympathetic nervous system development
          parasympathetic nervous system development
          inner ear development
          efferent axon development in a lateral line nerve
          respiratory system development
          retrotrapezoid nucleus neuron differentiation
          sympathetic ganglion development
          negative regulation of cell cycle arrest
          dopaminergic neuron differentiation
          cellular response to BMP stimulus
          neural crest cell migration involved in autonomic nervous system development
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding

          Back To Top

          Related Products

          • POP-6™ Polymer
          • POP-7™ Polymer
          • BigDye® Terminator v1.1 Cycle Sequencing Kit
          • BigDye® Terminator v3.1 Cycle Sequencing Kit
          • BigDye® Direct Cycle Sequencing Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline