Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FGFR1
Transcript:
NM_001174064.1
Assay Name:
Mutation genome location:
chr.8 38414721-38414867 on build GRCh38
Assay ID
Hs00522022_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
angiogenesis
ureteric bud development
in utero embryonic development
organ induction
neuron migration
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
transcription, DNA-templated
protein phosphorylation
sensory perception of sound
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
positive regulation of phospholipase C activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
cell migration
peptidyl-tyrosine phosphorylation
stem cell population maintenance
motogenic signaling involved in postnatal olfactory bulb interneuron migration
ventricular zone neuroblast division
embryonic limb morphogenesis
midbrain development
neuron projection development
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
inner ear morphogenesis
outer ear morphogenesis
middle ear morphogenesis
chordate embryonic development
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
regulation of cell differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of cell cycle
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
paraxial mesoderm development
regulation of lateral mesodermal cell fate specification
cell maturation
skeletal system morphogenesis
mesenchymal cell differentiation
regulation of sensory perception of pain
positive regulation of cardiac muscle cell proliferation
auditory receptor cell development
branching involved in salivary gland morphogenesis
lung-associated mesenchyme development
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
regulation of stem cell proliferation
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
regulation of extrinsic apoptotic signaling pathway in absence of ligand
glycoprotein binding
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein complex binding
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
cell adhesion molecule binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2w88v:80/100.66.78.250:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0