Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
BCL10
Transcript:
ENST00000370580.0
Assay Name:
Mutation genome location:
chr.1 85270731-85270884 on build GRCh38
Assay ID
Hs00522584_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Gene Ontology Categories:
Function(s)
Process(es)
B cell apoptotic process
neural tube closure
stimulatory C-type lectin receptor signaling pathway
toll-like receptor signaling pathway
response to molecule of bacterial origin
adaptive immune response
negative regulation of mature B cell apoptotic process
cellular defense response
I-kappaB kinase/NF-kappaB signaling
cell death
response to fungus
immunoglobulin mediated immune response
positive regulation of protein ubiquitination
response to food
positive regulation of mast cell cytokine production
Fc-epsilon receptor signaling pathway
lymphotoxin A biosynthetic process
interleukin-6 biosynthetic process
positive regulation of phosphorylation
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
innate immune response
positive regulation of interleukin-8 biosynthetic process
positive regulation of transcription, DNA-templated
T cell receptor signaling pathway
regulation of T cell receptor signaling pathway
positive regulation of T cell activation
positive regulation of NF-kappaB transcription factor activity
protein oligomerization
protein homooligomerization
protein heterooligomerization
T cell apoptotic process
cellular response to mechanical stimulus
positive regulation of extrinsic apoptotic signaling pathway
protease binding
transcription coactivator activity
protein binding
protein C-terminus binding
transcription factor binding
kinase activator activity
enzyme binding
kinase binding
protein kinase binding
ubiquitin protein ligase binding
identical protein binding
ubiquitin binding
protein kinase B binding
protein self-association
NF-kappaB binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline