Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ERCC3
Transcript:
NM_000122.1
Assay Name:
Mutation genome location:
chr.2 127257593-127257763 on build GRCh38
Assay ID
Hs00523498_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA helicase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
nucleotide-excision repair, DNA duplex unwinding
response to hypoxia
DNA topological change
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
protein phosphorylation
apoptotic process
response to oxidative stress
protein localization
response to UV
UV protection
viral process
nucleotide-excision repair, DNA incision
hair cell differentiation
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
embryonic organ development
global genome nucleotide-excision repair
regulation of mitotic cell cycle phase transition
DNA binding
damaged DNA binding
ATP-dependent DNA helicase activity
helicase activity
protein kinase activity
protein binding
ATP binding
GTP binding
protein C-terminus binding
DNA-dependent ATPase activity
transcription factor binding
RNA polymerase II carboxy-terminal domain kinase activity
ATPase activity
dATP binding
peptide binding
3'-5' DNA helicase activity
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline