Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NCOA1
Transcript:
NM_003743.4
Assay Name:
Mutation genome location:
chr.2 24658628-24658798 on build GRCh38
Assay ID
Hs00525401_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of transcription from RNA polymerase II promoter by galactose
regulation of thyroid hormone mediated signaling pathway
transcription, DNA-templated
lactation
male gonad development
bile acid and bile salt transport
cerebellum development
hippocampus development
hypothalamus development
cerebral cortex development
androgen receptor signaling pathway
intracellular receptor signaling pathway
response to estradiol
response to retinoic acid
response to progesterone
cellular response to hormone stimulus
positive regulation of apoptotic process
histone H4 acetylation
cellular lipid metabolic process
estrous cycle
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of female receptivity
positive regulation of transcription from RNA polymerase II promoter
male mating behavior
labyrinthine layer morphogenesis
cellular response to Thyroglobulin triiodothyronine
regulation of cellular response to drug
RNA polymerase II regulatory region DNA binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
ligand-dependent nuclear receptor binding
enzyme binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
protein complex binding
progesterone receptor binding
nuclear hormone receptor binding
retinoid X receptor binding
protein dimerization activity
protein N-terminus binding
androgen receptor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline