Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CSMD3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_052900.2 | NP_443132.3 | - | - | - |
NM_198123.1 | NP_937756.1 | - | - | - | |
NM_198124.1 | NP_937757.1 | 6111925 | c.10594G>T | p.E3532* | |
3951157 | c.10574C>A | p.A3525D | |||
7419480 | c.10539G>A | p.M3513I | |||
3644155 | c.10584G>A | p.K3528K | |||
1635586 | c.10603T>C | p.W3535R | |||
7208719 | c.10562T>G | p.V3521G | |||
7130847 | c.10552C>T | p.L3518F | |||
5437718 | c.10569T>G | p.A3523A | |||
1144773 | c.10561G>T | p.V3521L | |||
7363044 | c.10611G>C | p.M3537I | |||
3432032 | c.10602T>C | p.N3534N | |||
5048136 | c.10573G>A | p.A3525T | |||
1235995 | c.10579G>A | p.V3527M | |||
5039476 | c.10529C>T | p.A3510V | |||
6111921 | c.10520G>T | p.S3507I | |||
6111927 | c.10600A>T | p.N3534Y | |||
3644157 | c.10550A>C | p.Y3517S | |||
6410306 | c.10548A>G | p.I3516M | |||
3163186 | c.10534C>A | p.L3512I | |||
6492910 | c.10547delT | p.I3516fs*12 | |||
1202455 | c.10600A>G | p.N3534D | |||
6636680 | c.10565C>A | p.P3522H | |||
5520798 | c.10552C>G | p.L3518V | |||
4650063 | c.10543C>T | p.R3515C | |||
1454131 | c.10531C>T | p.R3511C | |||
1150866 | c.10589T>C | p.M3530T | |||
1238610 | c.10571A>G | p.H3524R | |||
1700479 | c.10594G>A | p.E3532K | |||
1597416 | c.10525G>T | p.E3509* | |||
5408657 | c.10525G>A | p.E3509K | |||
6111923 | c.10540T>C | p.L3514L | |||
XM_011516815.2 | XP_011515117.1 | - | - | - | |
XM_011516816.2 | XP_011515118.1 | - | - | - | |
XM_017013008.1 | XP_016868497.1 | - | - | - | |
XM_017013009.1 | XP_016868498.1 | 4650062 | c.10663C>T | p.R3555C | |
3644154 | c.10704G>A | p.K3568K | |||
1235994 | c.10699G>A | p.V3567M | |||
343760 | c.10732G>T | p.D3578Y | |||
1700478 | c.10714G>A | p.E3572K | |||
3163185 | c.10654C>A | p.L3552I | |||
5408656 | c.10645G>A | p.E3549K | |||
6111926 | c.10720A>T | p.N3574Y | |||
6111922 | c.10660T>C | p.L3554L | |||
5039475 | c.10649C>T | p.A3550V | |||
3432031 | c.10722T>C | p.N3574N | |||
5048135 | c.10693G>A | p.A3565T | |||
6636679 | c.10685C>A | p.P3562H | |||
5520797 | c.10672C>G | p.L3558V | |||
7363043 | c.10731G>C | p.M3577I | |||
748718 | c.10709T>C | p.M3570T | |||
7208718 | c.10682T>G | p.V3561G | |||
6492909 | c.10667delT | p.I3556fs*12 | |||
1202454 | c.10720A>G | p.N3574D | |||
5437717 | c.10689T>G | p.A3563A | |||
7419479 | c.10659G>A | p.M3553I | |||
1095041 | c.10645G>T | p.E3549* | |||
3644156 | c.10670A>C | p.Y3557S | |||
6410305 | c.10668A>G | p.I3556M | |||
25380 | c.10681G>T | p.V3561L | |||
6111924 | c.10714G>T | p.E3572* | |||
6111920 | c.10640G>T | p.S3547I | |||
3951156 | c.10694C>A | p.A3565D | |||
1454130 | c.10651C>T | p.R3551C | |||
1635585 | c.10723T>C | p.W3575R | |||
1238609 | c.10691A>G | p.H3564R | |||
7130846 | c.10672C>T | p.L3558F | |||
XM_017013010.1 | XP_016868499.1 | - | - | - | |
XM_017013011.1 | XP_016868500.1 | - | - | - | |
XM_017013012.1 | XP_016868501.1 | - | - | - | |
GenBank | AB067481.2 | BAB67787.2 | - | - | - |
AB114604.1 | BAC82443.1 | - | - | - | |
AB114605.1 | BAC82444.1 | - | - | - | |
AK095111.1 | - | - | - | ||
AY210419.1 | AAO34702.1 | - | - | - | |
ENST00000297405.0 | - | - | - |
Literature Links: |
CSMD3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |