Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PARP1
Transcript:
NM_001618.3
Assay Name:
Mutation genome location:
chr.1 226365814-226365987 on build GRCh38
Assay ID
Hs00531916_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
nucleotide-excision repair, DNA damage recognition
nucleotide-excision repair, DNA duplex unwinding
double-strand break repair via homologous recombination
lagging strand elongation
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
double-strand break repair
transcription from RNA polymerase II promoter
protein ADP-ribosylation
cellular response to DNA damage stimulus
mitochondrion organization
transforming growth factor beta receptor signaling pathway
positive regulation of cardiac muscle hypertrophy
protein autoprocessing
protein sumoylation
signal transduction involved in regulation of gene expression
macrophage differentiation
mitochondrial DNA metabolic process
cellular response to insulin stimulus
nucleotide-excision repair, DNA incision
cellular response to oxidative stress
protein modification process
DNA damage response, detection of DNA damage
mitochondrial DNA repair
positive regulation of transcription from RNA polymerase II promoter
DNA ligation involved in DNA repair
positive regulation of SMAD protein import into nucleus
protein poly-ADP-ribosylation
global genome nucleotide-excision repair
regulation of cellular protein localization
negative regulation of telomere maintenance via telomere lengthening
ATP generation from poly-ADP-D-ribose
positive regulation of transcription regulatory region DNA binding
DNA binding
DNA ligase (ATP) activity
NAD+ ADP-ribosyltransferase activity
protein binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
identical protein binding
histone deacetylase binding
poly(A) RNA binding
protein N-terminus binding
NAD binding
R-SMAD binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline