Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PCDHGA1
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA10
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA11
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA2
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA3
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA4
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA5
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA6
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA7
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA8
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGA9
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB1
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB2
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB3
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB4
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB5
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB6
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Gene:
PCDHGB7
Transcript:
NM_018914.2
Assay Name:
Mutation genome location:
chr.5 141422790-141423059 on build GRCh38
Assay transcription exon location:
Within Exon 1
Assay ID
Hs00546396_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
endosome to lysosome transport
lipid tube assembly involved in organelle fusion
cell adhesion
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
biological_process
calcium ion binding
poly(A) RNA binding
molecular_function
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline