Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MAP2K1
Transcript:
NM_002755.3
Assay Name:
Mutation genome location:
chr.15 66435094-66435319 on build GRCh38
Assay ID
Hs00564426_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
movement of cell or subcellular component
chemotaxis
cell cycle arrest
signal transduction
heart development
negative regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
peptidyl-tyrosine phosphorylation
cerebellar cortex formation
neuron differentiation
keratinocyte differentiation
thyroid gland development
regulation of stress-activated MAPK cascade
proteolysis in other organism
positive regulation of transcription, DNA-templated
thymus development
regulation of axon regeneration
cell motility
positive regulation of axonogenesis
Bergmann glial cell differentiation
face development
trachea formation
epithelial cell proliferation involved in lung morphogenesis
placenta blood vessel development
labyrinthine layer development
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of protein serine/threonine kinase activity
regulation of Golgi inheritance
cellular senescence
mitophagy in response to mitochondrial depolarization
positive regulation of production of miRNAs involved in gene silencing by miRNA
regulation of early endosome to late endosome transport
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
receptor signaling protein tyrosine phosphatase activity
protein binding
ATP binding
protein C-terminus binding
protein kinase binding
protein serine/threonine kinase activator activity
protein N-terminus binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit