Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TTN
Transcript:
NM_133437.4
Assay Name:
Mutation genome location:
chr.2 178564815-178565060 on build GRCh38
Assay transcription exon location:
Within Exon 155
Gene:
TTN-AS1
Transcript:
NM_133437.4
Assay Name:
Mutation genome location:
chr.2 178564815-178565060 on build GRCh38
Assay transcription exon location:
Within Exon 155
Assay ID
Hs00576778_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
cardiac muscle hypertrophy
muscle contraction
striated muscle contraction
mitotic chromosome condensation
peptidyl-tyrosine phosphorylation
muscle filament sliding
skeletal muscle thin filament assembly
skeletal muscle myosin thick filament assembly
detection of muscle stretch
sarcomere organization
regulation of protein kinase activity
cardiac muscle fiber development
sarcomerogenesis
regulation of catalytic activity
response to calcium ion
cardiac myofibril assembly
cardiac muscle tissue morphogenesis
cardiac muscle contraction
protease binding
protein serine/threonine kinase activity
protein tyrosine kinase activity
calcium ion binding
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
enzyme binding
protein kinase binding
telethonin binding
identical protein binding
actinin binding
protein self-association
actin filament binding
muscle alpha-actinin binding
structural molecule activity conferring elasticity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline