Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SLIT2
Transcript:
NM_004787.3
Assay Name:
Mutation genome location:
chr.4 20486089-20486350 on build GRCh38
Assay ID
Hs00577139_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
immunoglobulin superfamily cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
negative regulation of protein phosphorylation
cell migration involved in sprouting angiogenesis
negative regulation of leukocyte chemotaxis
axon guidance
motor neuron axon guidance
negative regulation of lamellipodium assembly
negative regulation of endothelial cell migration
negative regulation of smooth muscle cell migration
spinal cord development
chemorepulsion involved in embryonic olfactory bulb interneuron precursor migration
chemorepulsion involved in postnatal olfactory bulb interneuron migration
corticospinal neuron axon guidance through spinal cord
negative regulation of cell growth
negative regulation of cell migration
negative regulation of actin filament polymerization
retinal ganglion cell axon guidance
response to nutrient levels
cellular response to hormone stimulus
negative regulation of GTPase activity
Roundabout signaling pathway
positive regulation of apoptotic process
negative regulation of vascular permeability
branching morphogenesis of an epithelial tube
axon extension involved in axon guidance
negative regulation of inflammatory response
positive regulation of axonogenesis
negative chemotaxis
induction of negative chemotaxis
negative regulation of small GTPase mediated signal transduction
response to cortisol
apoptotic process involved in luteolysis
negative regulation of chemokine-mediated signaling pathway
cellular response to heparin
negative regulation of smooth muscle cell chemotaxis
negative regulation of mononuclear cell migration
negative regulation of neutrophil chemotaxis
negative regulation of monocyte chemotaxis
negative regulation of retinal ganglion cell axon guidance
negative regulation of cellular response to growth factor stimulus
GTPase inhibitor activity
calcium ion binding
protein binding
heparin binding
identical protein binding
protein homodimerization activity
laminin-1 binding
proteoglycan binding
heparan sulfate proteoglycan binding
Roundabout binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline