Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
IGF2
Transcript:
NM_001007139.5
Assay Name:
Mutation genome location:
chr.11 2135156-2135407 on build GRCh38
Gene:
INS-IGF2
Transcript:
NM_001007139.5
Assay Name:
Mutation genome location:
chr.11 2135156-2135407 on build GRCh38
Assay ID
Hs00590117_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
positive regulation of protein phosphorylation
platelet degranulation
glucose metabolic process
regulation of gene expression by genetic imprinting
regulation of transcription, DNA-templated
multicellular organism development
female pregnancy
memory
positive regulation of cell proliferation
insulin receptor signaling pathway
response to radiation
response to nutrient levels
response to estradiol
response to nicotine
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
wound healing
positive regulation of activated T cell proliferation
response to drug
positive regulation of catalytic activity
positive regulation of MAPK cascade
cellular protein metabolic process
response to ethanol
positive regulation of glycogen biosynthetic process
positive regulation of mitotic nuclear division
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of insulin receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
positive regulation of protein kinase B signaling
cellular response to mechanical stimulus
positive regulation of protein serine/threonine kinase activity
positive regulation of receptor activity
positive regulation of glycogen (starch) synthase activity
insulin receptor binding
insulin-like growth factor receptor binding
hormone activity
protein binding
growth factor activity
receptor activator activity
protein serine/threonine kinase activator activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline