Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NCKAP1L
Transcript:
NM_005337.4
Assay Name:
Mutation genome location:
chr.12 54523329-54523602 on build GRCh38
Assay ID
Hs00595980_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
B cell homeostasis
myeloid cell homeostasis
protein complex assembly
chemotaxis
maintenance of cell polarity
neutrophil chemotaxis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
positive regulation of B cell proliferation
activation of protein kinase activity
negative regulation of interleukin-17 production
negative regulation of interleukin-6 production
positive regulation of cell adhesion mediated by integrin
negative regulation of myosin-light-chain-phosphatase activity
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of T cell proliferation
positive regulation of phosphorylation
response to drug
T cell homeostasis
negative regulation of apoptotic process
positive regulation of CD4-positive, alpha-beta T cell differentiation
positive regulation of CD8-positive, alpha-beta T cell differentiation
positive regulation of GTPase activity
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
positive regulation of lymphocyte differentiation
positive regulation of erythrocyte differentiation
vascular endothelial growth factor receptor signaling pathway
erythrocyte development
B cell receptor signaling pathway
positive regulation of phagocytosis, engulfment
actin polymerization-dependent cell motility
positive regulation of neutrophil chemotaxis
GTPase activator activity
protein binding
protein kinase activator activity
protein complex binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline