Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TP53
Transcript:
NM_001126112.2
Assay Name:
Mutation genome location:
chr.17 7670484-7670753 on build GRCh38
Assay ID
Hs00600664_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline