Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
BRCA2
Transcript:
ENST00000380152.0
Assay Name:
Mutation genome location:
chr.13 32319158-32319383 on build GRCh38
Assay ID
Hs00638200_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
telomere maintenance via recombination
double-strand break repair via homologous recombination
DNA synthesis involved in DNA repair
strand displacement
cytokinesis
oocyte maturation
inner cell mass cell proliferation
nucleotide-excision repair
double-strand break repair
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
male meiosis I
spermatogenesis
brain development
cell aging
response to nutrient
female gonad development
response to X-ray
response to UV-C
response to gamma radiation
hemopoiesis
mammary gland development
chromosome breakage
homologous chromosome orientation involved in meiotic metaphase I plate congression
response to estradiol
regulation of cytokinesis
negative regulation of mammary gland epithelial cell proliferation
multicellular organism growth
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
histone H3 acetylation
histone H4 acetylation
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
replication fork protection
centrosome duplication
establishment of protein localization to telomere
mitotic recombination-dependent replication fork processing
protease binding
single-stranded DNA binding
histone acetyltransferase activity
protein binding
H3 histone acetyltransferase activity
H4 histone acetyltransferase activity
gamma-tubulin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline