Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ITGB1
Transcript:
NM_033668.2
Assay Name:
Mutation genome location:
chr.10 32911569-32911794 on build GRCh38
Assay ID
Hs00650041_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
in utero embryonic development
cell fate specification
tissue homeostasis
cell migration involved in sprouting angiogenesis
cellular calcium ion homeostasis
cellular defense response
homophilic cell adhesion via plasma membrane adhesion molecules
leukocyte cell-cell adhesion
cell-matrix adhesion
calcium-independent cell-matrix adhesion
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
germ cell migration
visual learning
regulation of collagen catabolic process
positive regulation of cell-substrate adhesion
positive regulation of neuron projection development
response to activity
cell migration
formation of radial glial scaffolds
B cell differentiation
extracellular matrix organization
positive regulation of cell migration
negative regulation of cell projection organization
cell-substrate adhesion
receptor internalization
protein transport within lipid bilayer
cell-cell adhesion mediated by integrin
heterotypic cell-cell adhesion
response to gonadotropin
negative regulation of Rho protein signal transduction
response to drug
positive regulation of apoptotic process
stress fiber assembly
positive regulation of MAPK cascade
positive regulation of GTPase activity
sarcomere organization
negative regulation of neuron differentiation
positive regulation of endocytosis
viral entry into host cell
mesodermal cell differentiation
axon extension
dendrite morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
regulation of immune response
leukocyte migration
leukocyte tethering or rolling
regulation of cell cycle
cardiac muscle cell differentiation
maternal process involved in female pregnancy
bicellular tight junction assembly
cellular response to mechanical stimulus
cellular response to vitamin D
cellular response to low-density lipoprotein particle stimulus
cellular response to ionizing radiation
positive regulation of establishment of protein localization to plasma membrane
negative regulation of anoikis
virus receptor activity
glycoprotein binding
fibronectin binding
protease binding
actin binding
integrin binding
protein binding
coreceptor activity
protein kinase binding
protein domain specific binding
C-X3-C chemokine binding
protein complex binding
peptide binding
laminin binding
metal ion binding
protein heterodimerization activity
cell adhesion molecule binding
alpha-actinin binding
collagen binding involved in cell-matrix adhesion
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline