Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RXRA
Transcript:
NM_002957.5
Assay Name:
Mutation genome location:
chr.9 134408721-134408990 on build GRCh38
Assay ID
Hs00655595_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
liver development
maternal placenta development
transcription initiation from RNA polymerase II promoter
vitamin metabolic process
inflammatory response
midgut development
embryo implantation
aging
cholesterol metabolic process
negative regulation of cell proliferation
response to selenium ion
bile acid and bile salt transport
modulation by virus of host morphology or physiology
axon regeneration
regulation of myelination
response to retinoic acid
cellular response to insulin stimulus
response to vitamin A
response to vitamin D
peroxisome proliferator activated receptor signaling pathway
camera-type eye development
positive regulation of apoptotic process
steroid hormone mediated signaling pathway
cellular lipid metabolic process
response to ethanol
positive regulation of transcription from RNA polymerase II promoter
positive regulation of translational initiation by iron
retinoic acid receptor signaling pathway
protein homotetramerization
response to glucocorticoid
ventricular cardiac muscle tissue morphogenesis
ventricular cardiac muscle cell differentiation
cardiac muscle cell proliferation
secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
regulation of branching involved in prostate gland morphogenesis
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
9-cis retinoic acid receptor activity
protein binding
zinc ion binding
enzyme binding
protein domain specific binding
chromatin DNA binding
protein complex binding
vitamin D receptor binding
retinoic acid receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
retinoic acid-responsive element binding
protein heterodimerization activity
vitamin D response element binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline