Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PRKCQ
Transcript:
NM_001323267.1
Assay Name:
Mutation genome location:
chr.10 6479069-6479304 on build GRCh38
Assay ID
Hs00659976_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
response to hypoxia
regulation of transcription, DNA-templated
membrane protein ectodomain proteolysis
cellular component disassembly involved in execution phase of apoptosis
inflammatory response
axon guidance
aging
response to heat
response to glucose
regulation of G2/M transition of mitotic cell cycle
response to organic cyclic compound
peptidyl-serine phosphorylation
regulation of vasoconstriction
platelet activation
positive regulation of telomere maintenance via telomerase
positive regulation of interleukin-17 production
positive regulation of interleukin-4 production
response to insulin
intracellular signal transduction
Fc-epsilon receptor signaling pathway
positive regulation of T cell proliferation
tissue regeneration
positive regulation of NF-kappaB import into nucleus
positive regulation of interleukin-2 biosynthetic process
respiratory burst
negative regulation of insulin receptor signaling pathway
positive regulation of protein secretion
T cell receptor signaling pathway
positive regulation of T cell activation
positive regulation of NF-kappaB transcription factor activity
positive regulation of filopodium assembly
positive regulation of stress fiber assembly
positive regulation of telomerase activity
cell chemotaxis
negative regulation of T cell apoptotic process
regulation of platelet aggregation
positive regulation of telomere capping
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 2 cell activation
protein kinase activity
protein serine/threonine kinase activity
protein kinase C activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline