Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PTK2B
Transcript:
NM_173174.2
Assay Name:
Mutation genome location:
chr.8 27430807-27431074 on build GRCh38
Assay ID
Hs00660537_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
oocyte maturation
response to hypoxia
positive regulation of cell-matrix adhesion
sprouting angiogenesis
adaptive immune response
marginal zone B cell differentiation
protein complex assembly
protein phosphorylation
apoptotic process
response to stress
cellular defense response
response to osmotic stress
signal transduction
cell surface receptor signaling pathway
signal complex assembly
epidermal growth factor receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
integrin-mediated signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of cell shape
response to mechanical stimulus
response to hormone
response to glucose
response to lithium ion
positive regulation of endothelial cell migration
negative regulation of muscle cell apoptotic process
regulation of cGMP-mediated signaling
regulation of macrophage chemotaxis
positive regulation of neuron projection development
glial cell proliferation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
positive regulation of cell growth
positive regulation of cell migration
negative regulation of bone mineralization
regulation of cGMP biosynthetic process
positive regulation of actin filament polymerization
neuron projection development
regulation of inositol trisphosphate biosynthetic process
tumor necrosis factor-mediated signaling pathway
ionotropic glutamate receptor signaling pathway
response to immobilization stress
peptidyl-tyrosine autophosphorylation
response to cocaine
response to drug
response to hydrogen peroxide
activation of Janus kinase activity
negative regulation of apoptotic process
stress fiber assembly
negative regulation of potassium ion transport
positive regulation of JUN kinase activity
negative regulation of neuron apoptotic process
blood vessel endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase activity
innate immune response
positive regulation of nitric oxide biosynthetic process
bone resorption
response to ethanol
negative regulation of myeloid cell differentiation
positive regulation of translation
positive regulation of angiogenesis
positive regulation of protein kinase activity
positive regulation of JNK cascade
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
focal adhesion assembly
positive regulation of peptidyl-tyrosine phosphorylation
regulation of calcium-mediated signaling
positive regulation of nitric-oxide synthase activity
regulation of release of sequestered calcium ion into cytosol
response to cAMP
response to calcium ion
positive regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
long term synaptic depression
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to retinoic acid
cellular response to fluid shear stress
activation of GTPase activity
regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
regulation of establishment of cell polarity
regulation of actin cytoskeleton reorganization
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of B cell chemotaxis
positive regulation of DNA biosynthetic process
calmodulin-dependent protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
NMDA glutamate receptor activity
receptor binding
protein binding
ATP binding
protein complex binding
3-phosphoinositide-dependent protein kinase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline