Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ATP1B1
Transcript:
ENST00000367816.0
Assay Name:
Mutation genome location:
chr.1 169131145-169131406 on build GRCh38
Assay ID
Hs00670850_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of defense response to virus by host
cellular calcium ion homeostasis
cellular sodium ion homeostasis
cell adhesion
potassium ion import
establishment or maintenance of transmembrane electrochemical gradient
regulation of gene expression
regulation of cardiac muscle contraction by calcium ion signaling
cellular potassium ion homeostasis
positive regulation of ATPase activity
ion transmembrane transport
sodium ion export from cell
protein transport into plasma membrane raft
ATP metabolic process
protein stabilization
leukocyte migration
relaxation of cardiac muscle
cardiac muscle contraction
protein localization to plasma membrane
membrane repolarization
membrane repolarization during cardiac muscle cell action potential
cell communication by electrical coupling involved in cardiac conduction
ATP hydrolysis coupled transmembrane transport
mitophagy in response to mitochondrial depolarization
xenophagy
positive regulation of potassium ion transmembrane transporter activity
positive regulation of sodium ion export from cell
positive regulation of calcium:sodium antiporter activity
positive regulation of potassium ion import
regulation of cardiac conduction
potassium ion import across plasma membrane
ATPase activator activity
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
protein C-terminus binding
drug binding
ATPase activity
MHC class II protein complex binding
potassium ion binding
sodium ion binding
ATPase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline