Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
INHBA
Transcript:
NM_002192.2
Assay Name:
Mutation genome location:
chr.7 41690018-41690277 on build GRCh38
Assay transcription exon location:
Within Exon 3
Assay ID
Hs00674593_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
ovarian follicle development
hair follicle development
hematopoietic progenitor cell differentiation
regulation of transcription from RNA polymerase II promoter
defense response
cell cycle arrest
cell surface receptor signaling pathway
cell-cell signaling
nervous system development
negative regulation of cell proliferation
male gonad development
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
striatal medium spiny neuron differentiation
cell differentiation
erythrocyte differentiation
negative regulation of cell growth
positive regulation of cellular protein metabolic process
activin receptor signaling pathway
endodermal cell differentiation
growth
negative regulation of phosphorylation
odontogenesis
response to drug
hemoglobin biosynthetic process
progesterone secretion
regulation of MAPK cascade
negative regulation of interferon-gamma biosynthetic process
negative regulation of B cell differentiation
positive regulation of erythrocyte differentiation
negative regulation of macrophage differentiation
negative regulation of cell cycle
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of follicle-stimulating hormone secretion
positive regulation of follicle-stimulating hormone secretion
negative regulation of follicle-stimulating hormone secretion
mesodermal cell differentiation
cell development
palate development
positive regulation of ovulation
SMAD protein signal transduction
eyelid development in camera-type eye
cellular response to follicle-stimulating hormone stimulus
cellular response to cholesterol
GABAergic neuron differentiation
extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
transforming growth factor beta receptor binding
hormone activity
protein binding
growth factor activity
peptide hormone binding
inhibin binding
identical protein binding
protein heterodimerization activity
type II activin receptor binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline