Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
XBP1
Transcript:
NM_005080.3
Assay Name:
Mutation genome location:
chr.22 28798970-28799243 on build GRCh38
Assay ID
Hs00684873_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
liver development
endothelial cell proliferation
positive regulation of immunoglobulin production
transcription from RNA polymerase II promoter
ubiquitin-dependent protein catabolic process
fatty acid biosynthetic process
autophagy
immune response
positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
muscle organ development
regulation of autophagy
negative regulation of myotube differentiation
protein transport
endoplasmic reticulum unfolded protein response
exocrine pancreas development
protein destabilization
cellular response to nutrient
cellular response to insulin stimulus
cellular triglyceride homeostasis
IRE1-mediated unfolded protein response
ATF6-mediated unfolded protein response
cellular response to glucose starvation
cholesterol homeostasis
negative regulation of apoptotic process
positive regulation of MHC class II biosynthetic process
positive regulation of B cell differentiation
positive regulation of T cell differentiation
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
neuron development
positive regulation of immunoglobulin secretion
fatty acid homeostasis
sterol homeostasis
adipose tissue development
epithelial cell maturation involved in salivary gland development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to fructose stimulus
cellular response to glucose stimulus
cellular response to interleukin-4
cellular response to peptide hormone stimulus
positive regulation of plasma cell differentiation
positive regulation of endoplasmic reticulum unfolded protein response
positive regulation of proteasomal protein catabolic process
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of lactation
response to insulin-like growth factor stimulus
positive regulation of hepatocyte proliferation
positive regulation of interleukin-6 secretion
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
RNA polymerase II transcription factor binding
enhancer sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
phosphatidylinositol 3-kinase regulatory subunit binding
histone deacetylase binding
protein heterodimerization activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline