Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NOD2
Transcript:
NM_022162.2
Assay Name:
Mutation genome location:
chr.16 50699834-50700089 on build GRCh38
Assay ID
Hs00705967_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cytokine production involved in immune response
cytokine secretion involved in immune response
positive regulation of dendritic cell antigen processing and presentation
positive regulation of type 2 immune response
defense response
JNK cascade
detection of biotic stimulus
detection of bacterium
maintenance of gastrointestinal epithelium
response to muramyl dipeptide
detection of muramyl dipeptide
positive regulation of interleukin-1 beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-17 production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
positive regulation of stress-activated MAPK cascade
activation of MAPK activity involved in innate immune response
intracellular signal transduction
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatidylinositol 3-kinase activity
innate immune response
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of gamma-delta T cell activation
positive regulation of epithelial cell proliferation
positive regulation of interleukin-1 beta secretion
regulation of inflammatory response
positive regulation of B cell activation
positive regulation of NF-kappaB transcription factor activity
protein oligomerization
positive regulation of oxidoreductase activity
positive regulation of nitric-oxide synthase biosynthetic process
positive regulation of prostaglandin-endoperoxide synthase activity
positive regulation of ERK1 and ERK2 cascade
nucleotide-binding oligomerization domain containing signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
cellular response to muramyl dipeptide
positive regulation of cytokine production involved in inflammatory response
positive regulation of NIK/NF-kappaB signaling
negative regulation of macrophage apoptotic process
positive regulation of prostaglandin-E synthase activity
actin binding
protein binding
ATP binding
enzyme binding
protein kinase binding
Hsp70 protein binding
muramyl dipeptide binding
peptidoglycan binding
CARD domain binding
Hsp90 protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline