Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LGALS9
Transcript:
NM_009587.2
Assay Name:
Mutation genome location:
chr.17 27646442-27646711 on build GRCh38
Assay ID
Hs00721643_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
extracellular matrix protein
Gene Ontology Categories:
Function(s)
Process(es)
natural killer cell tolerance induction
chemotaxis
inflammatory response
female pregnancy
positive regulation of gene expression
negative regulation of gene expression
response to lipopolysaccharide
regulation of interleukin-4 production
regulation of interleukin-5 production
negative regulation of chemokine production
negative regulation of interferon-gamma production
negative regulation of tumor necrosis factor production
positive regulation of interleukin-4 production
positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation involved in immune response
toll-like receptor 2 signaling pathway
toll-like receptor 4 signaling pathway
p38MAPK cascade
positive regulation of NF-kappaB import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of mast cell degranulation
negative regulation of natural killer cell mediated cytotoxicity
negative regulation of activated T cell proliferation
positive regulation of viral entry into host cell
positive regulation of interleukin-1 beta secretion
positive regulation of NF-kappaB transcription factor activity
maternal process involved in female pregnancy
positive regulation of activated T cell autonomous cell death
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
response to interleukin-1
cellular response to interferon-gamma
positive regulation of transforming growth factor beta production
positive regulation of monocyte chemotactic protein-1 production
mature conventional dendritic cell differentiation
cellular response to virus
regulation of p38MAPK cascade
positive regulation of interferon-gamma secretion
positive regulation of tumor necrosis factor secretion
positive regulation of interleukin-8 secretion
positive regulation of dendritic cell chemotaxis
negative regulation of CD4-positive, alpha-beta T cell proliferation
positive regulation of CD4-positive, alpha-beta T cell proliferation
positive regulation of interleukin-13 secretion
positive regulation of dendritic cell apoptotic process
positive regulation of interleukin-6 secretion
positive regulation of interleukin-10 secretion
positive regulation of interleukin-12 secretion
positive regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
positive regulation of dendritic cell differentiation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
signal transducer activity
galactose binding
enzyme binding
carbohydrate binding
disaccharide binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline