Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
DNMT1
Transcript:
NM_001379.3
Assay Name:
Mutation genome location:
chr.19 10137611-10137868 on build GRCh38
Assay ID
Hs00727837_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA methyltransferase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
transcription, DNA-templated
Ras protein signal transduction
brain development
aging
response to heat
response to toxic substance
response to ionizing radiation
maintenance of DNA methylation
DNA methylation on cytosine within a CG sequence
positive regulation of gene expression
response to activity
gene silencing
covalent chromatin modification
neuron differentiation
response to caffeine
response to estradiol
response to lipopolysaccharide
response to vitamin A
response to testosterone
cellular response to platelet-derived growth factor stimulus
response to antidepressant
regulation of cell proliferation
DNA hypermethylation
response to ethanol
negative regulation of gene expression, epigenetic
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
S-adenosylmethionine metabolic process
positive regulation of histone H3-K4 methylation
negative regulation of histone H3-K9 methylation
cellular response to amino acid stimulus
cellular response to lead ion
cellular response to transforming growth factor beta stimulus
C-5 methylation of cytosine
positive regulation of methylation-dependent chromatin silencing
cellular response to nerve growth factor stimulus
DNA binding
double-stranded DNA binding
RNA binding
DNA (cytosine-5-)-methyltransferase activity
protein binding
zinc ion binding
methyl-CpG binding
DNA-methyltransferase activity
protein domain specific binding
estrogen receptor binding
histone deacetylase binding
unmethylated CpG binding
DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates
promoter-specific chromatin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline