Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PTPN6
Transcript:
NM_080549.3
Assay Name:
Mutation genome location:
chr.12 6955397-6955668 on build GRCh38
Assay transcription exon location:
Overlaps Exon 6 - Exon 7
Assay ID
Hs00730532_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
negative regulation of humoral immune response mediated by circulating immunoglobulin
protein dephosphorylation
apoptotic process
G-protein coupled receptor signaling pathway
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cell differentiation
platelet activation
platelet formation
T cell costimulation
abortive mitotic cell cycle
positive regulation of cell adhesion mediated by integrin
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
megakaryocyte development
negative regulation of T cell proliferation
natural killer cell mediated cytotoxicity
negative regulation of MAP kinase activity
regulation of B cell differentiation
negative regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
leukocyte migration
regulation of release of sequestered calcium ion into cytosol
regulation of type I interferon-mediated signaling pathway
regulation of ERK1 and ERK2 cascade
platelet aggregation
regulation of G1/S transition of mitotic cell cycle
phosphotyrosine binding
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
SH3 domain binding
protein kinase binding
SH2 domain binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline