Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Gene:
          PCDHGA1
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA10
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA11
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA12
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA2
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA3
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA4
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA5
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA6
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA7
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA8
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGA9
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB1
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB2
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB3
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB4
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB5
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB6
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Gene:
          PCDHGB7
          Transcript:
          NM_003735.2
          Assay Name:
          Mutation genome location:
          chr.5 141431032-141431293 on build GRCh38
          Assay transcription exon location:
          Within Exon 1
          Assay ID Hs00760357_CE

          Purification
          Size
          Availability Made To Order
          Catalog # A15629
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details


          Back To Top

          More Information


          Set Membership:

          Between 200-400 bp

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          endosome to lysosome transport
          lipid tube assembly involved in organelle fusion
          cell adhesion
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          biological_process
          calcium ion binding
          poly(A) RNA binding
          molecular_function

          Back To Top

          Related Products

          • POP-6™ Polymer
          • POP-7™ Polymer
          • BigDye® Terminator v1.1 Cycle Sequencing Kit
          • BigDye® Terminator v3.1 Cycle Sequencing Kit
          • BigDye® Direct Cycle Sequencing Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline