Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ILK
Transcript:
NM_006284.3
Assay Name:
Mutation genome location:
chr.11 6610783-6611008 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 5
Gene:
TAF10
Transcript:
NM_006284.3
Assay Name:
Mutation genome location:
chr.11 6610783-6611008 on build GRCh38
Assay transcription exon location:
Overlaps - Exon 5
Assay ID
Hs00774620_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
positive regulation of cell-matrix adhesion
outflow tract morphogenesis
protein phosphorylation
negative regulation of protein kinase activity
cell cycle arrest
cell-matrix adhesion
integrin-mediated signaling pathway
cell aging
cell proliferation
positive regulation of cell proliferation
negative regulation of cardiac muscle cell apoptotic process
fibroblast migration
negative regulation of smooth muscle cell migration
peptidyl-serine phosphorylation
nerve development
myelination in peripheral nervous system
positive regulation of cell migration
positive regulation of BMP signaling pathway
myelin assembly
regulation of actin cytoskeleton organization
cell junction assembly
substrate adhesion-dependent cell spreading
positive regulation of phosphorylation
extracellular fibril organization
positive regulation of MAP kinase activity
protein kinase B signaling
negative regulation of neuron apoptotic process
establishment or maintenance of epithelial cell apical/basal polarity
positive regulation of myoblast differentiation
positive regulation of osteoblast differentiation
positive regulation of axon extension
positive regulation of transcription, DNA-templated
negative regulation of smooth muscle cell proliferation
neuron projection morphogenesis
positive regulation of dendrite morphogenesis
protein heterooligomerization
positive regulation of protein kinase B signaling
platelet aggregation
positive regulation of canonical Wnt signaling pathway
negative regulation of neural precursor cell proliferation
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone deubiquitination
histone H3 acetylation
protein homooligomerization
regulation of signal transduction by p53 class mediator
protein serine/threonine kinase activity
signal transducer activity
integrin binding
protein binding
ATP binding
SH3 domain binding
protein kinase binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
enzyme binding
estrogen receptor binding
RNA polymerase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline